Gene Transcripts
- Gene Symbol
- Tgfb1i1
- Entrez Gene ID
- 21804
- Gene Name
- transforming growth factor beta 1 induced transcript 1
- Gene Aliases
- ARA55, Hic5, TSC-5, hic-5
- Chromosome Location
- Chr. 7: 128246799 - 128255699 on Build GRCm38
- UniGene ID
- Mm.3248
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289550.1 |
NP_001276479.1
|
4, 5 |
501 |
|
NM_001289551.1 |
NP_001276480.1
|
4, 5 |
501 |
|
NM_001289553.1 |
NP_001276482.1
|
3, 4 |
385 |
|
GenBank mRNA |
AK003170.1 |
- |
4, 5 |
336 |
|
AK041233.1 |
- |
4, 5 |
329 |
|
AK085042.1 |
- |
3, 4 |
227 |
|
AK087114.1 |
- |
4, 5 |
336 |
|
AK156423.1 |
- |
4, 5 |
336 |
|
BC002049.1 |
- |
4, 5 |
335 |
3489459 |
BC056362.1 |
- |
3, 4 |
1610 |
6816930 |
DQ143891.1 |
- |
3, 4 |
386 |
|
DQ143892.1 |
- |
2, 3 |
201 |
|
DQ143895.1 |
- |
2, 3 |
590 |
|
DQ143898.1 |
- |
3, 4 |
463 |
|
DQ143899.1 |
- |
2, 3 |
590 |
|
DQ143900.1 |
- |
3, 4 |
398 |
|
L22482.1 |
- |
3, 4 |
462 |
|
View Full Product Details