Gene Transcripts

Gene Symbol
Wnt9b
Entrez Gene ID
22412
Gene Name
wingless-type MMTV integration site family, member 9B
Gene Aliases
Wnt14b, Wnt15, clf, clf1, wnt-14b, wnt-15
Chromosome Location
Chr. 11: 103727363 - 103749821 on Build GRCm38
UniGene ID
Mm.215161
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011719.4 NP_035849.3 4 783
GenBank mRNA AB073819.1 - 4 746
AF469004.1 - 4 731
AK052708.1 - 4 784
BC110482.1 - 4 775 40048674

View Full Product Details