Gene Transcripts

Gene Symbol
Wt1
Entrez Gene ID
22431
Gene Name
Wilms tumor 1 homolog
Gene Aliases
D630046I19Rik, Wt-1
Chromosome Location
Chr. 2: 105126529 - 105173614 on Build GRCm38
UniGene ID
Mm.389339
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144783.2 NP_659032.3 3, 4 1146
GenBank mRNA DQ537939.1 - 3, 4 483
M55512.1 - 3, 4 1151

View Full Product Details