Gene Transcripts

Gene Symbol
Xpa
Entrez Gene ID
22590
Gene Name
xeroderma pigmentosum, complementation group A
Gene Aliases
AI573865, Xpac
Chromosome Location
Chr. 4: 46174746 - 46196349 on Build GRCm38
UniGene ID
Mm.247036
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011728.2 NP_035858.2 6 791
GenBank mRNA AF354052.1 - 4 413
AK133577.1 - 1 2688
X74351.1 - 6 791

View Full Product Details