Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001045863.1 | NP_001039328.1 | 4 | 448 | |
NM_001045864.1 | NP_001039329.1 | 4 | 534 | ||
NM_138580.2 | NP_613046.2 | 4 | 322 | ||
GenBank mRNA | AK011663.1 | - | 4 | 306 | |
AK012389.1 | - | 4 | 339 | ||
AK131947.1 | - | 4 | 306 | ||
AK147011.1 | - | 4 | 430 | ||
BC025905.1 | - | 1 | 488 | ||
BC034867.1 | - | 4 | 274 | 1495547 | |
CN525174.1 | - | 4 | 277 | ||
DV662511.1 | - | 1 | 67 |