Gene Transcripts

Gene Symbol
Diap2
Entrez Gene ID
54004
Gene Name
diaphanous homolog 2 (Drosophila)
Gene Aliases
Dia3, Diaph2, Drf2, E430022I22Rik
Chromosome Location
Chr. X: 129749668 - 130465834 on Build GRCm38
UniGene ID
Mm.10763
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172493.2 NP_766081.1 23 2986
GenBank mRNA AB713187.1 - 24 2781
AK083254.1 - 16 1918
AK088648.1 - 23 2920
AK153986.1 - 23 2974
AY312280.1 - 23 2923
Y15910.1 - 4 493

View Full Product Details