Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016867.1 | NP_058563.1 | 3 | 588 | |
GenBank mRNA | AF061262.1 | - | 3 | 588 | |
AK008307.1 | - | 3 | 405 | ||
AK008626.1 | - | 3 | 601 | ||
AK168560.1 | - | 3 | 599 | ||
BC013440.1 | - | 3 | 642 | 4238716 |