Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018777.4 | NP_061247.1 | 3 | 937 | |
GenBank mRNA | AK010560.1 | - | 3 | 938 | |
AK010682.1 | - | 3 | 932 | ||
BC005718.1 | - | 3 | 914 | 3594823 | |
BC050138.1 | - | 3 | 920 | 30028814 |