Gene Transcripts

Gene Symbol
Ppie
Entrez Gene ID
56031
Gene Name
peptidylprolyl isomerase E (cyclophilin E)
Gene Aliases
2010010D16Rik, Cyp33
Chromosome Location
Chr. 4: 123127114 - 123140000 on Build GRCm38
UniGene ID
Mm.126873
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019489.5 NP_062362.1 2 77
GenBank mRNA AF182825.1 - 2 59
AK008177.1 - 2 78
AK042165.1 - 1 1027
BC045154.1 - 2 72 6468589

View Full Product Details