Gene Transcripts

Gene Symbol
Cyp3a25
Entrez Gene ID
56388
Gene Name
cytochrome P450, family 3, subfamily a, polypeptide 25
Gene Aliases
AI327008
Chromosome Location
Chr. 5: 145977194 - 146009639 on Build GRCm38
UniGene ID
Mm.301900
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019792.2 NP_062766.2 11 1200
GenBank mRNA AF204959.1 - 11 1199
AK149541.1 - 10 1113
BC028855.1 - 11 1191 5124151
Y11995.1 - 11 1202

Gene Symbol
Cyp3a59
Entrez Gene ID
100041449
Gene Name
cytochrome P450, family 3, subfamily a, polypeptide 59
Gene Aliases
ENSMUSG00000061292
Chromosome Location
Chr. 5: 146079209 - 146113791 on Build GRCm38
UniGene ID
Mm.301900
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105160.1 NP_001098630.1 11 1200

View Full Product Details