Gene Transcripts

Gene Symbol
Cenpk
Entrez Gene ID
60411
Gene Name
centromere protein K
Gene Aliases
B130045K24Rik, BB232189, C530004N04Rik, Cenp-K, Solt, Solzt
Chromosome Location
Chr. 13: 104228611 - 104249622 on Build GRCm38
UniGene ID
Mm.281498
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021790.2 NP_068562.1 2 333
NM_181061.5 NP_851406.1 2 456
GenBank mRNA AB043687.1 - 2 333
AK033117.1 - 1 1828
AK082939.1 - 2 113
AK160776.1 - 2 91
BC028655.1 - 2 456
BC092351.1 - 2 95

View Full Product Details