Gene Transcripts

Gene Symbol
Spcs2
Entrez Gene ID
66624
Gene Name
signal peptidase complex subunit 2 homolog (S. cerevisiae)
Gene Aliases
5730406I15Rik, AA408713, mKIAA0102
Chromosome Location
Chr. 7: 99837569 - 99870341 on Build GRCm38
UniGene ID
Mm.30043
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025668.3 NP_079944.1 5 583
GenBank mRNA AK014310.1 - 5 597
AK017504.1 - 5 588
AK077510.1 - 5 593
AK134578.1 - 5 584
AK168173.1 - 5 588
AK188156.1 - 1 25
BC010547.1 - 4 401
BC050150.1 - 5 593 30026746

View Full Product Details