Gene Transcripts

Gene Symbol
Eif4e3
Entrez Gene ID
66892
Gene Name
eukaryotic translation initiation factor 4E member 3
Gene Aliases
1300018P11Rik, AI451927, eIF4E-3
Chromosome Location
Chr. 6: 99625137 - 99666771 on Build GRCm38
UniGene ID
Mm.32889
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025829.4 NP_080105.1 3 276
GenBank mRNA AK005054.2 - 3 277
AK007268.1 - 3 265
AY628329.1 - 3 257
BC027014.1 - 3 271 5369891

View Full Product Details