Gene Transcripts
- Gene Symbol
- Lrrc40
- Entrez Gene ID
- 67144
- Gene Name
- leucine rich repeat containing 40
- Gene Aliases
- 2610040E16Rik, AI837674
- Chromosome Location
- Chr. 3: 158036682 - 158067090 on Build GRCm38
- UniGene ID
- Mm.358704
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289524.1 |
NP_001276453.1
|
6, 7 |
853 |
|
NM_001289525.1 |
NP_001276454.1
|
4, 5 |
600 |
|
NM_024194.6 |
NP_077156.2
|
6, 7 |
856 |
|
GenBank mRNA |
AK011726.1 |
- |
6, 7 |
837 |
|
AK017814.1 |
- |
6, 7 |
825 |
|
AK034978.1 |
- |
|
836 |
|
AK148748.1 |
- |
6, 7 |
816 |
|
AK152244.1 |
- |
4, 5 |
581 |
|
AK163288.1 |
- |
6, 7 |
824 |
|
AK170052.1 |
- |
6, 7 |
833 |
|
BC002279.1 |
- |
2, 3 |
245 |
3497400 |
View Full Product Details