Gene Transcripts
- Gene Symbol
- Cylc1
- Entrez Gene ID
- 67407
- Gene Name
- cylicin, basic protein of sperm head cytoskeleton 1
- Gene Aliases
- 4930401M13Rik
- Chromosome Location
- Chr. X: 111110418 - 111125961 on Build GRCm38
- UniGene ID
- Mm.426142
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026134.2 |
NP_080410.2
|
3, 4 |
162 |
|
GenBank mRNA |
AK015303.1 |
- |
3, 4 |
164 |
|
View Full Product Details