Gene Transcripts
- Gene Symbol
- Slco6b1
- Entrez Gene ID
- 67854
- Gene Name
- solute carrier organic anion transporter family, member 6b1
- Gene Aliases
- 1700022M03Rik
- Chromosome Location
- Chr. 1: 96906176 - 96997560 on Build GRCm38
- UniGene ID
- Mm.263706
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK006249.1 |
- |
4 |
454 |
|
AK016463.1 |
- |
6 |
1200 |
|
AY461596.1 |
- |
6 |
1206 |
|
View Full Product Details