Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012328.2 | NP_036460.1 | 2 | 533 | |
GenBank mRNA | AB026908.1 | - | 2 | 366 | |
AF083247.1 | - | 2 | 367 | ||
AF093824.1 | - | 2 | 333 | ||
AK092204.1 | - | 3 | 391 | ||
AL080081.1 | - | 2 | 413 | ||
AY359045.1 | - | 1 | 187 | ||
BC028912.1 | - | 2 | 347 | 4943933 | |
CR533475.1 | - | 1 | 164 |