Gene Transcripts
- Gene Symbol
- Leprotl1
- Entrez Gene ID
- 68192
- Gene Name
- leptin receptor overlapping transcript-like 1
- Gene Aliases
- 1110067H13Rik, 1520402O14Rik, AI854296
- Chromosome Location
- Chr. 8: 34135572 - 34146739 on Build GRCm38
- UniGene ID
- Mm.34212
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026609.2 |
NP_080885.1
|
2, 3 |
126 |
|
GenBank mRNA |
AK003329.1 |
- |
2, 3 |
137 |
|
AK013870.1 |
- |
2, 3 |
196 |
|
AK075740.1 |
- |
2, 3 |
181 |
|
AK150289.1 |
- |
2, 3 |
185 |
|
AK153277.1 |
- |
2, 3 |
171 |
|
AK154712.1 |
- |
2, 3 |
191 |
|
AK159401.1 |
- |
2, 3 |
114 |
|
AK203783.1 |
- |
1, 2 |
83 |
|
BC004677.1 |
- |
2, 3 |
126 |
3500223 |
View Full Product Details