Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001024205.2 | NP_001019376.1 | 2 | 628 | |
GenBank mRNA | AK030113.1 | - | 2 | 630 | |
AK122494.1 | - | 2 | 612 | ||
AK132603.1 | - | 596 | |||
AK142954.1 | - | 2 | 598 | ||
AK152336.1 | - | 2 | 599 | ||
AK160601.1 | - | 2 | 629 | ||
AK170764.1 | - | 2 | 626 |