Gene Transcripts

Gene Symbol
Lamtor5
Entrez Gene ID
68576
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 5
Gene Aliases
1110003H18Rik, Hbxip, XIP
Chromosome Location
Chr. 3: 107278858 - 107284082 on Build GRCm38
UniGene ID
Mm.290035
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026774.2 NP_081050.2 1 228
GenBank mRNA AI552736.1 - 1 228
AK003358.1 - 1 68
AK146289.1 - 1 88
BC028547.1 - 1 58 1513442

View Full Product Details