Gene Transcripts

Gene Symbol
Dopey2
Entrez Gene ID
70028
Gene Name
dopey family member 2
Gene Aliases
0610038M01Rik, 2610510B01Rik, AI503246, C21orf5, mKIAA0933
Chromosome Location
Chr. 16: 93711907 - 93810588 on Build GRCm38
UniGene ID
Mm.23230
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026700.2 NP_080976.1 2, 3 323
NM_027293.1 NP_081569.1 2, 3 323
GenBank mRNA AK002808.1 - 2, 3 253
AK147255.1 - 2, 3 301
AK156170.1 - 2, 3 297
AK164839.1 - 2, 3 300
BY195296.1 - 2, 3 323

View Full Product Details