Gene Transcripts

Gene Symbol
Prss22
Entrez Gene ID
70835
Gene Name
protease, serine 22
Gene Aliases
4733401N09Rik, BSSP-4, SP001LA
Chromosome Location
Chr. 17: 23993534 - 24000735 on Build GRCm38
UniGene ID
Mm.157351
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133731.2 NP_598492.2 6 955
GenBank mRNA AB010778.1 - 6 935
AK014645.1 - 6 955
BC055854.1 - 6 940 4035351

View Full Product Details