Gene Transcripts

Gene Symbol
R3hcc1
Entrez Gene ID
71843
Gene Name
R3H domain and coiled-coil containing 1
Gene Aliases
1700020M16Rik, AA407796
Chromosome Location
Chr. 14: 69697304 - 69714809 on Build GRCm38
UniGene ID
Mm.27738
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146012.2 NP_001139484.2 5 1116
NM_001301651.1 NP_001288580.1 5 1068
NM_001301652.1 NP_001288581.1 4 1349
GenBank mRNA AK006177.1 - 5 1068
AK032854.1 - 5 1086
AK156843.1 - 5 1086
BC052536.1 - 4 1265
BC059860.1 - 4 1349

View Full Product Details