Gene Transcripts

Gene Symbol
Pygo1
Entrez Gene ID
72135
Gene Name
pygopus 1
Gene Aliases
2600014C22Rik
Chromosome Location
Chr. 9: 72925650 - 72946015 on Build GRCm38
UniGene ID
Mm.273605
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028116.2 NP_082392.1 3 696
GenBank mRNA AK011208.1 - 3 698
BC116630.1 - 3 533 40057035
BC116631.1 - 3 533 40057038

View Full Product Details