Gene Transcripts

Gene Symbol
Ms4a6c
Entrez Gene ID
73656
Gene Name
membrane-spanning 4-domains, subfamily A, member 6C
Gene Aliases
2200009H22Rik, 2210417N07Rik
Chromosome Location
Chr. 19: 11469041 - 11487069 on Build GRCm38
UniGene ID
Mm.440719
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028595.4 NP_082871.2 5 672
GenBank mRNA AF237910.1 - 5 630
AK008652.1 - 5 646
AK150397.1 - 5 569
AK152258.1 - 5 569

View Full Product Details