Gene Transcripts

Gene Symbol
Prss51
Entrez Gene ID
100504162
Gene Name
protease, serine 51
Gene Aliases
1700007N14Rik
Chromosome Location
Chr. 14: 64085525 - 64097674 on Build GRCm38
UniGene ID
Mm.72752
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193631.1 NP_001180560.1 4 619
GenBank mRNA AK005740.1 - 4 624
BC048657.1 - 4 619

View Full Product Details