Gene Transcripts

Gene Symbol
NAP1L2
Entrez Gene ID
4674
Gene Name
nucleosome assembly protein 1 like 2
Gene Aliases
BPX
Chromosome Location
Chr. X: 73212299 - 73214874 on Build GRCh38
UniGene ID
Hs.66180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021963.3 NP_068798.1 1 1148
GenBank mRNA AB027013.1 - 1 1122
AF136178.1 - 1 1096
AK303676.1 - 2 741
AK316569.1 - 1 1148
BC026325.1 - 1 1070 4798964
CR457095.1 - 1 766

View Full Product Details