Gene Transcripts

Gene Symbol
Pelp1
Entrez Gene ID
75273
Gene Name
proline, glutamic acid and leucine rich protein 1
Gene Aliases
4930563C04Rik, MNAR
Chromosome Location
Chr. 11: 70392881 - 70410031 on Build GRCm38
UniGene ID
Mm.340601
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029231.4 NP_083507.3 1 127
GenBank mRNA AK005027.1 - 1 125
AK016197.1 - 1 128
AK128966.1 - 1 127
BC090620.1 - 1 121 30699968

View Full Product Details