Gene Transcripts

Gene Symbol
Krtap9-3
Entrez Gene ID
75586
Gene Name
keratin associated protein 9-3
Gene Aliases
2310040M23Rik
Chromosome Location
Chr. 11: 99597348 - 99598106 on Build GRCm38
UniGene ID
Mm.75707
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029351.2 NP_083627.2 1 672
GenBank mRNA AK009730.1 - 1 674
AK131927.1 - 1 673

View Full Product Details