Gene Transcripts
- Gene Symbol
- NDUFS6
- Entrez Gene ID
- 4726
- Gene Name
- NADH:ubiquinone oxidoreductase subunit S6
- Gene Aliases
- CI-13kA, CI-13kD-A, CI13KDA
- Chromosome Location
- Chr. 5: 1801382 - 1816051 on Build GRCh38
- UniGene ID
- Hs.408257
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004553.4 |
NP_004544.1
|
1, 2 |
175 |
|
GenBank mRNA |
AF044959.1 |
- |
1, 2 |
149 |
|
AK022503.1 |
- |
1, 2 |
158 |
|
BC038664.1 |
- |
1, 2 |
154 |
5204222 |
BC046155.2 |
- |
1, 2 |
144 |
6047460 |
BG706812.1 |
- |
1, 2 |
164 |
|
BM172493.1 |
- |
1, 2 |
30 |
|
BM759028.1 |
- |
1, 2 |
162 |
|
BM764340.1 |
- |
1, 2 |
162 |
|
CN285188.1 |
- |
1, 2 |
129 |
|
CR456880.1 |
- |
1, 2 |
139 |
|
View Full Product Details