Gene Transcripts
- Gene Symbol
- Ppp1r14c
- Entrez Gene ID
- 76142
- Gene Name
- protein phosphatase 1, regulatory (inhibitor) subunit 14c
- Gene Aliases
- 6330514J04Rik, KEPI
- Chromosome Location
- Chr. 10: 3366071 - 3464979 on Build GRCm38
- UniGene ID
- Mm.308126
- Species
- Mus musculus
- Gene Symbol
- Gm14057
- Entrez Gene ID
- 100043766
- Gene Name
- Ppp1r14c pseudogene
- Gene Aliases
- OTTMUSG00000015563
- Chromosome Location
- Chr. 2: 130801966 - 130804101 on Build GRCm38
- UniGene ID
- Mm.447040
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK013448.1 |
- |
1 |
243 |
|
BC057040.1 |
- |
|
2068 |
|
View Full Product Details