Gene Transcripts

Gene Symbol
Mmab
Entrez Gene ID
77697
Gene Name
methylmalonic aciduria (cobalamin deficiency) type B homolog (human)
Gene Aliases
9130222L19Rik, ATR
Chromosome Location
Chr. 5: 114431034 - 114444073 on Build GRCm38
UniGene ID
Mm.105182
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029956.3 NP_084232.1 9 677
GenBank mRNA AK020286.1 - 9 677
AK088785.1 - 9 669
AK148421.1 - 9 712
BC022159.1 - 9 631
BC057558.1 - 9 677 6415979

View Full Product Details