Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145510.1 | NP_663485.1 | 2 | 343 | |
GenBank mRNA | AK087547.1 | - | 2 | 338 | |
AK133528.1 | - | 2 | 364 | ||
AK155054.1 | - | 2 | 343 | ||
AK156126.1 | - | 255 | |||
AK158129.1 | - | 356 | |||
AK165748.1 | - | 2 | 361 | ||
BC011166.1 | - | 2 | 343 | 4206804 | |
BC024808.1 | - | 2 | 333 | 5356768 |