Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133973.2 | NP_598734.1 | 10 | 1288 | |
GenBank mRNA | AK156466.1 | - | 10 | 1279 | |
AK172465.1 | - | 10 | 1279 | ||
AK191447.1 | - | 2 | 126 | ||
AK199943.1 | - | 2 | 126 | ||
AY223866.1 | - | 10 | 1315 | ||
BC023120.1 | - | 10 | 1265 | 4161858 |