Gene Transcripts

Gene Symbol
OMD
Entrez Gene ID
4958
Gene Name
osteomodulin
Gene Aliases
OSAD, SLRR2C
Chromosome Location
Chr. 9: 92414245 - 92424554 on Build GRCh38
UniGene ID
Hs.94070
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005014.2 NP_005005.1 2 501
GenBank mRNA AB000114.1 - 2 232
AK225922.1 - 2 346
AK313055.1 - 2 340
AY358872.1 - 2 399
BC046356.1 - 2 403 5239983

View Full Product Details