Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024233.3 | NP_077195.2 | 2 | 270 | |
GenBank mRNA | AK007731.1 | - | 2 | 272 | |
AK090202.1 | - | 2 | 274 | ||
AK150287.1 | - | 2 | 186 | ||
AK171619.1 | - | 2 | 173 | ||
AK172641.1 | - | 2 | 248 | ||
AK178028.1 | - | 2 | 112 | ||
AK209299.1 | - | 2 | 112 | ||
AK211686.1 | - | 2 | 159 | ||
AK212689.1 | - | 2 | 159 | ||
BC003445.1 | - | 2 | 206 | 2651110 |