Gene Transcripts
- Gene Symbol
- ACYP2
- Entrez Gene ID
- 98
- Gene Name
- acylphosphatase 2
- Gene Aliases
- ACYM, ACYP
- Chromosome Location
- Chr. 2: 53970838 - 54305300 on Build GRCh38
- UniGene ID
- Hs.516173
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320586.1 |
NP_001307515.1
|
5, 6 |
548 |
|
NM_001320587.1 |
NP_001307516.1
|
4, 5 |
455 |
|
NM_001320588.1 |
NP_001307517.1
|
2, 3 |
249 |
|
NM_001320589.1 |
NP_001307518.1
|
2, 3 |
500 |
|
NM_001320590.1 |
NP_001307519.1
|
2, 3 |
500 |
|
NM_138448.3 |
NP_612457.1
|
2, 3 |
500 |
|
XM_017005406.1 |
XP_016860895.1
|
6, 7 |
894 |
|
XM_017005407.1 |
XP_016860896.1
|
5, 6 |
813 |
|
XM_017005408.1 |
XP_016860897.1
|
4, 5 |
1522 |
|
XM_017005409.1 |
XP_016860898.1
|
4, 5 |
720 |
|
XM_017005410.1 |
XP_016860899.1
|
2, 3 |
370 |
|
XM_017005411.1 |
XP_016860900.1
|
6, 7 |
894 |
|
XM_017005412.1 |
XP_016860901.1
|
6, 7 |
894 |
|
XM_017005413.1 |
XP_016860902.1
|
6, 7 |
894 |
|
GenBank mRNA |
AK307096.1 |
- |
2, 3 |
371 |
|
BC012290.1 |
- |
2, 3 |
269 |
3864319 |
BF671963.1 |
- |
2, 3 |
248 |
|
BG741807.1 |
- |
4, 5 |
449 |
|
BU543943.1 |
- |
5, 6 |
548 |
|
BU857820.1 |
- |
2, 3 |
358 |
|
X84195.1 |
- |
2, 3 |
249 |
|
View Full Product Details