Gene Transcripts

Gene Symbol
Tas2r116
Entrez Gene ID
112408
Gene Name
taste receptor, type 2, member 116
Gene Aliases
T2R16, TRB1, TRB4, Tas2r14, Tas2r16, Tas2r7, mGR16, mt2r56
Chromosome Location
Chr. 6: 132855438 - 132856355 on Build GRCm38
UniGene ID
Mm.261566
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053212.1 NP_444442.1 1 228
GenBank mRNA AF247734.1 - 1 135

View Full Product Details