Gene Transcripts

Gene Symbol
Des
Entrez Gene ID
64362
Gene Name
desmin
Gene Aliases
Chromosome Location
Chr. 9: 82556574 - 82564288 on Build Rnor_6.0
UniGene ID
Rn.39196
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022531.1 NP_071976.1 2 668
GenBank mRNA BC061872.1 - 2 661

View Full Product Details