Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZIGA4
Entrez Gene ID
837390
Gene Name
ARF GAP-like zinc finger-containing protein ZIGA4
Gene Aliases
AGD14, ARF GAP-like zinc finger-containing protein ZIGA4, ARF-GAP domain 14, AT1G08680, AT1G08690, F22O13.16, F22O13_16
Chromosome Location
Chr.1: 2762584 - 2768967 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035925.2 NP_001031002.1 1 - 2 333 70
NM_001198013.1 NP_001184942.1 1 - 2 339 70
NM_001331785.1 NP_001320735.1 1 - 2 333 70
NM_100741.4 NP_172344.2 1 - 2 333 70
NM_180610.4 NP_850941.1 1 - 2 334 70
GenBank mRNA AF184145.1 - 1 - 2 256 70
AK176415.1 - 1 - 2 280 70
AY081303.1 - 1 - 2 317 70
BT008398.1 - 1 - 2 103 70

View Full Product Details