Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLS1
Entrez Gene ID
830765
Gene Name
flavonol synthase 1
Gene Aliases
AT5G08640, ATFLS1, FLAVONOL SYNTHASE, FLS, T2K12.5, flavonol synthase 1
Chromosome Location
Chr.5: 2803833 - 2805606 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203337.1 NP_001190266.1 1 - 2 518 89
NM_120951.3 NP_196481.1 1 - 2 642 89
GenBank mRNA AY058068.1 - 1 - 2 512 89
AY086328.1 - 1 - 2 518 89
BT000494.1 - 1 - 2 466 89
U84259.1 - 1 - 2 512 89
U84260.1 - 1 - 2 512 89

View Full Product Details