Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCV2
Entrez Gene ID
840912
Gene Name
like COV 2
Gene Aliases
AT1G43130, F1I21.3, F1I21_3, like COV 2
Chromosome Location
Chr.1: 16228189 - 16231254 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333208.1 NP_001321430.1 3 - 4 450 88
NM_001333209.1 NP_001321431.1 2 - 3 383 88
NM_103466.5 NP_564483.1 2 - 3 270 88
GenBank mRNA AK176692.1 - 2 - 3 249 88
AK176760.1 - 2 - 3 249 88
BT033158.1 - 2 - 3 174 88

View Full Product Details