Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
iqd17
Entrez Gene ID
828002
Gene Name
IQ-domain 17
Gene Aliases
AT4G00820, A_TM018A10.13, A_TM018A10_13, IQ-domain 17, T18A10.11, T18A10_11
Chromosome Location
Chr.4: 349025 - 351550 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116308.4 NP_567191.2 4 - 5 1097 123

View Full Product Details