Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TET10
Entrez Gene ID
842632
Gene Name
tetraspanin10
Gene Aliases
AT1G63260, F9N12.12, F9N12_12, tetraspanin10
Chromosome Location
Chr.1: 23466700 - 23469289 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036145.3 NP_001031222.1 3 - 4 553 77
NM_001334075.1 NP_001320678.1 3 - 4 530 77
NM_001334076.1 NP_001320677.1 3 - 4 572 77
NM_001334077.1 NP_001320676.1 4 - 5 471 77
NM_105005.5 NP_176515.3 3 - 4 553 77
NM_202348.3 NP_974077.1 3 - 4 553 77
GenBank mRNA BT011569.1 - 3 - 4 414 77
BT012253.1 - 3 - 4 228 77

View Full Product Details