Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KAT2
Entrez Gene ID
827555
Gene Name
potassium channel KAT1-like protein
Gene Aliases
AT4G18290, T9A21.140, T9A21_140, VOLTAGE-GATED POTASSIUM CHANNEL 2, potassium channel in Arabidopsis thaliana 2
Chromosome Location
Chr.4: 10114794 - 10118652 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341273.1 NP_001329801.1 9 - 10 2108 109
NM_117939.5 NP_193563.3 9 - 10 1484 109

View Full Product Details