Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DR4
Entrez Gene ID
843668
Gene Name
drought-repressed 4
Gene Aliases
AT1G73330, ATDR4, T9L24.45, T9L24_45, drought-repressed 4
Chromosome Location
Chr.1: 27568714 - 27569823 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105993.3 NP_177476.1 1 - 1 983 126

View Full Product Details