Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARF9
Entrez Gene ID
828498
Gene Name
auxin response factor 9
Gene Aliases
AT4G23980, T32A16.150, T32A16_150, auxin response factor 9
Chromosome Location
Chr.4: 12451143 - 12455369 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036629.2 NP_001031706.1 7 - 8 1004 114
NM_118530.5 NP_194129.1 7 - 8 1059 114
GenBank mRNA AF082176.1 - 7 - 8 897 114
AF344312.1 - 7 - 8 610 114

View Full Product Details