The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202989.1 | NP_001189918.1 | 2 - 3 | 221 | 117 |
NM_001338294.1 | NP_001326663.1 | 2 - 3 | 273 | 117 | |
NM_112639.4 | NP_566581.1 | 2 - 3 | 273 | 117 | |
GenBank mRNA | AK117162.1 | - | 2 - 3 | 215 | 117 |
BT024620.1 | - | 2 - 3 | 140 | 117 |