Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
U2A'
Entrez Gene ID
837505
Gene Name
U2 small nuclear ribonucleoprotein A
Gene Aliases
AT1G09760, F21M12.14, F21M12_14, U2 small nuclear ribonucleoprotein A
Chromosome Location
Chr.1: 3159125 - 3161692 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100848.4 NP_172447.1 1 - 2 172 68
GenBank mRNA AY059902.1 - 1 - 2 135 68
BT000143.1 - 1 - 2 83 68

View Full Product Details