Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COBL6
Entrez Gene ID
837508
Gene Name
COBRA-like protein 6 precursor
Gene Aliases
AT1G09790, COBRA-like protein 6 precursor, F21M12.17, F21M12_17
Chromosome Location
Chr.1: 3168434 - 3171018 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331851.1 NP_001322459.1 2 - 3 346 109
NM_100851.2 NP_172450.2 2 - 3 180 109

View Full Product Details